Order Entry
Poland
ContactUsLinkComponent
Anti-Cadherin 20 Rabbit Polyclonal Antibody
Anti-Cadherin 20 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13716R
Supplier:  Bioss
Anti-Cadherin 20 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13716R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin 20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    CDH7L3|Cdh7|Cadherin 20|FLJ37047.|Cadherin 20,cadherin-20|type 2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-20, also known as protocadherin gamma B4, is a member of the protocadherin gamma gene cluster. Expressed primariy in fibroblasts, cadherin-20 plays a critical role in neuronal cell-cell connections in the brain. Through its ability to mediate cell-cell adhesion, caderin-20 is thought to help the body respond to injury by regulating post-traumatic wound healing.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Cadherin 20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat