Order Entry
Poland
ContactUsLinkComponent
Potassium hydrogen monopersulphate for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Potassium hydrogen monopersulphate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Potassium hydrogen monopersulphate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Hill Formula:
    H₃K₅O₁₈S₄
  • Chemical formula:
    2KHSO₅ * KHSO₄ * K₂SO₄
  • Molar Mass:
    614.76 g/mol
  • Density:
    1.1 - 1.2 g/cm³ (20 °C)
  • pH value:
    2 - 3 (10 g/l, H₂O, 20 °C)
  • Solubility:
    256 g/l (20 °C)
  • LD 50 oral:
    LD50 Rat 2000 mg/kg
  • Storage class:
    8 B Non-combustible corrosive substances
  • WGK:
    WGK 1 slightly water endangering
  • R Phrase:
    R 8-34-37
  • S Phrase:
    S 17-26-36/37/39-45

Specification Test Results

Related Information

Frequently Bought Together