Anti-MCM8 Rabbit Polyclonal Antibody
Catalog # BSBTPB9722
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:minichromosome maintenance 8 homologous recombination repair factor
- Antigen symbol:MCM8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84515
- Antigen synonyms:dJ967N21.5|POF10|C20orf154
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for DNA helicase MCM8(MCM8) detection. Tested with WB in Human.
Type: Primary
Antigen: MCM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human