Order Entry
Poland
ContactUsLinkComponent
 
Anti-LIMK1 Rabbit Polyclonal Antibody
  BSBTPB9716
undefined
Anti-LIMK1 Rabbit Polyclonal Antibody
  BSBTPB9716

 

  • Antibody type:
    Primary
  • Antigen name:
    LIM domain kinase 1
  • Antigen symbol:
    LIMK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3984
  • Antigen synonyms:
    LIMK-1|LIMK
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for LIM domain kinase 1(LIMK1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: LIMK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat