Anti-ATG8 Rabbit Polyclonal Antibody
AGRIAS142769
: Agrisera
- Antibody type:Primary
- Antigen symbol:ATG8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Plant,C. reinhardtii,Synechocystis sp. pcc 6803,Cyanobacteria
- Western blot:Yes
- UniProtKB:Q8S925
- Immunogen:fragment of recombinant ATG8 from Chlamydomonas reinhardtii,
- Size:50 µl
- Pk:50 µl
ATG8 (Autophagy-related protein) involved in degradation and recycling of intracellular components in a process of autophagy. ATG8 is a molecular autophagy marker in Chlamydomonas reinhardtii (Pérez-Pérez et al. 2010, Plant Physiol. 152: 1874-88).,ATG8 (Autophagy-related protein) involved in degradation and recycling of intracellular components in a process of autophagy. ATG8 is a molecular autophagy marker in Chlamydomonas reinhardtii (Pérez-Pérez et al. 2010, Plant Physiol. 152: 1874-88).
Type: Primary
Antigen: ATG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, Cyanobacteria, Synechocystis sp. pcc 6803, Plants