Anti-PSA2 Rabbit Polyclonal Antibody
Catalog # AGRIAS132654
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- UniProtKB:O64750
- Immunogen:recombinant protein corresponding to amino acids 87 to 186 of Arabidopsis thaliana PSA2,
- Size:50 µl
- Pk:50 µl
Specifications
About this item
PSA2 (Photosystem I assembly factor 2) is a protein coded by a gene belonging to the "Green Cut" set, found only in green algae and plants but not in non-photosynthetic organisms. PSA2 is localized in thylakoid lumen.,PSA2 (Photosystem I assembly factor 2) is a protein coded by a gene belonging to the "Green Cut" set, found only in green algae and plants but not in non-photosynthetic organisms. PSA2 is localized in thylakoid lumen.
Type: Primary
Antigen: PSA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human