Order Entry
Poland
ContactUsLinkComponent
pH/mV/°C meters, handheld, FiveGo™ F2
pH/mV/°C meters, handheld, FiveGo™ F2
Supplier:  Mettler - Toledo
Certificates
undefined
pH/mV/°C meters, handheld, FiveGo™ F2
Supplier:  Mettler - Toledo

Specifications

  • Catalog No:
  • 662-2007
  • 662-2008
  • 662-2009
  • 662-2010
  • Model:
  • F2
  • F2
  • F2
  • F2
  • pH range:
  • 0,00 - 14,00
  • 0,00 - 14,00
  • 0,00 - 14,00
  • 0,00 - 14,00
  • pH resolution:
  • 0,01
  • 0,01
  • 0,01
  • 0,01
  • pH accuracy:
  • ±0,01
  • ±0,01
  • ±0,01
  • ±0,01
  • pH calibration:
  • 3 points (4 pre-defined buffer groups)
  • 3 points (4 pre-defined buffer groups)
  • 3 points (4 pre-defined buffer groups)
  • 3 points (4 pre-defined buffer groups)
  • mV range:
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • mV resolution:
  • 1
  • 1
  • 1
  • 1
  • mV accuracy:
  • ±1
  • ±1
  • ±1
  • ±1
  • Temperature range:
  • 0...+100 °C
  • 0...+100 °C
  • 0...+100 °C
  • 0...+100 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,5 °C
  • ±0,5 °C
  • ±0,5 °C
  • ±0,5 °C
  • Temperature compensation:
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • ATC & MTC
  • Display:
  • 3,1" segmented LCD
  • 3,1" segmented LCD
  • 3,1" segmented LCD
  • 3,1" segmented LCD
  • Power supply:
  • 4× AAA batteries
  • 4× AAA batteries
  • 4× AAA batteries
  • 4× AAA batteries
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Data logging capacity:
  • 200 measurements
  • 200 measurements
  • 200 measurements
  • 200 measurements
  • Connections:
  • BNC, Cinch, NTC 30 kΩ
  • BNC, Cinch, NTC 30 kΩ
  • BNC, Cinch, NTC 30 kΩ
  • BNC, Cinch, NTC 30 kΩ
  • W×D×H:
  • 188×77×33 mm
  • 188×77×33 mm
  • 188×77×33 mm
  • 188×77×33 mm
  • Weight:
  • 0,26 kg
  • 0,26 kg
  • 0,26 kg
  • 0,26 kg

Specifications

Related Information

Electrodes

Product Information

Frequently Bought Together