Order Entry
Poland
ContactUsLinkComponent
Anti-CD8a, CD11b/c, CD45R/B220, Ly-6G (Gr-1), MIHC Class II (I-A/I-E) Monoclonal Antibody (PE-Cy7®, APC-Cy7®, PerCP-Cy5.5®, APC, PE, FITC) [clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-85C, TER-119]
Anti-CD8a, CD11b/c, CD45R/B220, Ly-6G (Gr-1), MIHC Class II (I-A/I-E) Monoclonal Antibody (PE-Cy7®, APC-Cy7®, PerCP-Cy5.5®, APC, PE, FITC) [clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-85C, TER-119]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD8a, CD11b/c, CD45R/B220, Ly-6G (Gr-1), MIHC Class II (I-A/I-E) Monoclonal Antibody (PE-Cy7®, APC-Cy7®, PerCP-Cy5.5®, APC, PE, FITC) [clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-85C, TER-119]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 91-0004-U025
  • 91-0004-U100
  • Antigen symbol:
  • CD8A
  • CD8A
  • Antigen name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®/APC (Allophycocyanin)-Cy7®/PerCP-Cy5.5®/APC (Allophycocyanin)/PE (Phycoerythrin)/FITC (Fluorescein Isothiocyanate)
  • PE (Phycoerythrin)-Cy7®/APC (Allophycocyanin)-Cy7®/PerCP-Cy5.5®/APC (Allophycocyanin)/PE (Phycoerythrin)/FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-85C, TER-119
  • 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-85C, TER-119
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Multiple
  • Multiple
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together