Order Entry
Poland
ContactUsLinkComponent
Anti-CD23 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: EBVCS-5]
Anti-CD23 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: EBVCS-5]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD23 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: EBVCS-5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0237-T025
  • 20-0237-T500
  • 20-0237-T100
  • Antigen symbol:
  • FCER2
  • FCER2
  • FCER2
  • Antigen name:
  • Fc fragment of IgE receptor 2
  • Fc fragment of IgE receptor 2
  • Fc fragment of IgE receptor 2
  • Antigen synonyms:
  • CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
  • CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
  • CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • EBVCS-5
  • EBVCS-5
  • EBVCS-5
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 2208
  • 2208
  • 2208
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together