Anti-Lamin B1 Antibody
BSBTPB9611
- Antibody type:Primary
- Antigen name:lamin B1
- Antigen symbol:LMNB1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4001
- Antigen synonyms:ADLD|LMN2|LMNB|LMN
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Lamin-B1(LMNB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Lamin B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat