Single film frame shippers
Supplier: ENTEGRIS
Certificates
About this item
Film frame shippers provide a convenient, safe and ergonomic means to ship and store wafers mounted on film frames. Film frame shippers are designed to protect the frame and to prevent any contact with the wafer surface.
- Trays stack and snap together conveniently
- Multiple trays can be shipped or stored together for greater economy
- Lightweight (1 kg) and easy to carry
- Parabolic design prevents wafer contact
- Finger slots for safe wafer extraction
- Available in ESD protective materials
- RFID capable to enable monitoring/tracking of product
Frequently Bought Together