Anti-RNF181 Mouse Monoclonal Antibody
ABNOH00051255-PW1
: Abnova
- Antigen name:Ring Finger Protein 181
- Antigen symbol:RNF181
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:51255
- Antigen synonyms:HSPC238
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-RNF181 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-RNF181 (50 ?g)
Type:
Antigen: RNF181
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human