Anti-WAPAL Mouse Monoclonal Antibody
ABNOH00023063-PW2
: Abnova
- Nazwa antygenu:Wings Apart-Like
- Symbol antygenu:WAPAL
- Klonalność:Monoklonalne
- Gospodarz:Mouse
- Immunoprecypitacja:Yes
- Reaktywność:Human
- Western blot:Yes
- Identyfikator genu:23063
- Synonimy antygenów:FOE|Wings apart-like protein homolog|KIAA0261|WAPL|Friend of EBNA2 protein
- Temperatura przechowywania:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Rozmiar:1 Set
- Ilość:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-WAPAL (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-WAPAL (50 ?g)
Type:
Antigen: WAPAL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human