Order Entry
Poland
ContactUsLinkComponent
Anti-F11R Polyclonal Antibody Pair
Anti-F11R Polyclonal Antibody Pair
Catalog # ABNOH00050848-PW2
Supplier:  Abnova
Anti-F11R Polyclonal Antibody Pair
Catalog # ABNOH00050848-PW2
Supplier:  Abnova

Specifications

  • Antigen name:
    F11 receptor
  • Antigen symbol:
    F11R
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    50848
  • Antigen synonyms:
    JAMA|CD321|JCAM|PAM-1|KAT|JAM1|JAM
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-F11R (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-F11R (50 µg)

Type:
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human