Anti-COASY Polyclonal Antibody Pair
Catalog # ABNOH00080347-AP22
Supplier: Abnova
Specifications
- Antigen name:CoA synthase
- Antigen symbol:COASY
- Clonality:Polyclonal
- Host:Multiple
- Reactivity:Human
- Gene ID:80347
- Antigen synonyms:pOV-2|PPAT|UKR1|NBIA6|DPCK|NBP
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human COASY.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-COASY (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-COASY (20 µg);
Type:
Antigen: COASY
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human