Anti-EFHD1 Polyclonal Antibody Pair
ABNOH00080303-PW1
: Abnova
- Nazwa antygenu:EF-hand Domain Family, member D1
- Symbol antygenu:EFHD1
- Klonalność:Poliklonalne
- Gospodarz:Multiple
- Immunoprecypitacja:Yes
- Reaktywność:Human
- Western blot:Yes
- Identyfikator genu:80303
- Synonimy antygenów:MST133|MSTP133|EF-hand domain-containing protein D1|SWS2|PP3051|EF-hand domain-containing protein 1
- Temperatura przechowywania:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Rozmiar:1 Set
- Ilość:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-EFHD1 (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-EFHD1 (50 µl)
Type:
Antigen: EFHD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human