Anti-PLK4 Polyclonal Antibody Pair
ABNOH00010733-PW2
: Abnova
- Antigen name:Polo-like kinase 4
- Antigen symbol:PLK4
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:10733
- Antigen synonyms:STK18|MCCRP2|SAK
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-PLK4 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-PLK4 (50 µg)
Type:
Antigen: PLK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human