Order Entry
Poland
ContactUsLinkComponent
Anti-RARS Polyclonal Antibody Pair
Anti-RARS Polyclonal Antibody Pair
Catalog # ABNOH00005917-PW1
Supplier:  Abnova
Anti-RARS Polyclonal Antibody Pair
Catalog # ABNOH00005917-PW1
Supplier:  Abnova

Specifications

  • Antigen name:
    Arginyl-tRNA Synthetase
  • Antigen symbol:
    RARS
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    5917
  • Antigen synonyms:
    ArgRS, DALRD1, HLD9
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-RARS (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-RARS (50 µg)

Type:
Antigen: RARS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human