Anti-KLF15 Antibody Pair
Catalog # ABNOH00028999-PW1
Supplier: Abnova
Specifications
- Antigen name:Kruppel-like Factor 15
- Antigen symbol:KLF15
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:28999
- Antigen synonyms:DKFZp779M1320|KKLF
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-KLF15 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-KLF15 (50 µl)
Type:
Antigen: KLF15
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human