Anti-GSTA1 Rabbit Polyclonal Antibody
1 Set
Catalog # ABNOH00002938-PW1
Supplier: Abnova
Jump to:
Product Details & Documents
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-GSTA1 (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-GSTA1 (50 ?l)
Type:
Antigen: GSTA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Size:1 Set
- Western blot:Yes
- Cat. No.:ABNOH00002938-PW1
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Gene ID:2938
- Antigen synonyms:GSTA1-1|GTH1|GST2
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Host:Rabbit
- Antigen symbol:GSTA1
- Antigen name:Glutathione S-transferase alpha 1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



