Order Entry
Poland
ContactUsLinkComponent
Anti-CACNB3 Polyclonal Antibody Pair
Anti-CACNB3 Polyclonal Antibody Pair
Catalog # ABNOH00000784-PW1
Dostawca:  Abnova
Anti-CACNB3 Polyclonal Antibody Pair
Catalog # ABNOH00000784-PW1
Dostawca:  Abnova

Dane techniczne

  • Antigen name:
    Calcium Channel, Voltage-dependent, Beta 3 Subunit
  • Antigen symbol:
    CACNB3
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    784
  • Antigen synonyms:
    CACNLB3|CAB3
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Dane techniczne

Informacje o tym produkcie

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-CACNB3 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-CACNB3 (50 µg)

Type:
Antigen: CACNB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human