Anti-APRT Polyclonal Antibody Pair
Catalog # ABNOH00000353-PW1
Dostawca: Abnova
Dane techniczne
- Antigen name:Adenine Phosphoribosyltransferase
- Antigen symbol:APRT
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:353
- Antigen synonyms:AMP|APRTD
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Dane techniczne
Informacje o tym produkcie
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-APRT (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-APRT (50 µg)
Type:
Antigen: APRT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human