Anti-MLST8 Polyclonal Antibody Pair
ABNOH00064223-PW1
: Abnova
- Nazwa antygenu:MTOR associated protein, LST8 homolog
- Symbol antygenu:MLST8
- Klonalność:Poliklonalne
- Gospodarz:Multiple
- Immunoprecypitacja:Yes
- Reaktywność:Human
- Western blot:Yes
- Identyfikator genu:64223
- Synonimy antygenów:POP3|GbetaL|GBL|LST8|WAT1
- Temperatura przechowywania:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Rozmiar:1 Set
- Ilość:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-GBL (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-MLST8 (50 µl)
Type:
Antigen: MLST8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human