Anti-STK36 + GLI3 Antibody Pair
Catalog # ABNODI0590
Dostawca: Abnova
Dane techniczne
- Antigen symbol:STK36 + GLI3
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:27148|2737
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Dane techniczne
Informacje o tym produkcie
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the STK36 protein, and the other against the GLI3 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. STK36 Rabbit purified Polyclonal Antibody (20 µg) 2. GLI3 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: STK36 + GLI3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: