Anti-AKT1 + PPM1A Antibody Pair
ABNODI0384
: Abnova
- Antigen name:AKT1 + PPM1A
- Antigen symbol:AKT1 + PPM1A
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:207|5494
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the AKT1 protein, and the other against the PPM1A protein for use in Proximity Ligation Assay
Antibody pair set content: 1. AKT1 Rabbit purified Polyclonal Antibody (20 µg) 2. PPM1A Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: AKT1 + PPM1A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: