Anti-MAPK3 + RALGDS Antibody Pair
Catalog # ABNODI0265
Supplier: Abnova
Specifications
- Symbol antygenu:MAPK3 + RALGDS
- Gospodarz:Multiple
- ImmunoFluorescencja:Yes
- Identyfikator genu:5595|5900
- Temperatura przechowywania:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Rozmiar:1 Set
- Ilość:1 SET
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK3 protein, and the other against the RALGDS protein for use in Proximity Ligation Assay
Antibody pair set content: 1. MAPK3 Rabbit purified Polyclonal Antibody (20 µg) 2. RALGDS Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: MAPK3 + RALGDS
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: