Order Entry
Poland
ContactUsLinkComponent
Anti-APOA2 Antibody Pair
Anti-APOA2 Antibody Pair
Catalog # ABNOH00000336-AP11
Supplier:  Abnova
Anti-APOA2 Antibody Pair
Catalog # ABNOH00000336-AP11
Supplier:  Abnova

Specifications

  • Antigen name:
    Apolipoprotein A2
  • Antigen symbol:
    APOA2
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    336
  • Antigen synonyms:
    Apoa-2|ApoAII|Apo-AII|ApoA-II|Apolipoprotein A-II|Hdl-1|Alp-2|Apolipoprotein A2|Truncated Apolipoprotein A-II|Apolipoprotein A-II(1-76)
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human APOA2.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-APOA2 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-APOA2, IgG1 kappa (20 µg);

Type:
Antigen: APOA2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human