Order Entry
Poland
ContactUsLinkComponent
 
Anti-PRKCD + PRKCD Antibody Pair
  ABNODP0231
 :  Abnova
Anti-PRKCD + PRKCD Antibody Pair
  ABNODP0231
 :  Abnova

 

  • Antigen name:
    Protein Kinase C Delta
  • Antigen symbol:
    PRKCD + PRKCD
  • Host:
    Multiple
  • ImmunoFluorescence:
    Yes
  • Gene ID:
    5580
  • Antigen synonyms:
    CVID9|MAY1|nPKC-delta|ALPS3|PKCD
  • Modification:
    phospho-Ser645
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PRKCD protein, and the other against the specific S645 phosphorylated site of PRKCD for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PRKCD S645 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PRKCD Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PRKCD + PRKCD
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: