Order Entry
Poland
Orders LinkContactUsLinkComponent
Dissolution vessels, QLA
Dissolution vessels, QLA
Dissolution vessels, QLA
Certificates
Dissolution vessels, QLA
Dissolution vessels, QLA
3 Options Available Below
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Product Family Options
Glass
Product Information
  • Availability
    Price
  • QLAAGLA900-01
    QLA clear glass dissolution vessel, serialised, 1000 ml
    QLA clear glass dissolution vessel, serialised, 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 630 zł
    1 630 zł
    1 * 1 SZT
       1 630 zł
  • QLAAGLA900-HR
    QLA clear glass dissolution vessel, 1000 ml, Hanson compatible
    QLA clear glass dissolution vessel, 1000 ml, Hanson compatible
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 540 zł
    1 540 zł
    1 * 1 SZT
       1 540 zł
  • QLAAGLA900-VK
    QLA clear glass dissolution vessel, TruCenter, serialised, 1000 ml
    QLA clear glass dissolution vessel, TruCenter, serialised, 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 270 zł
    2 270 zł
    1 * 1 SZT
       2 270 zł