Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Chromatography Standards
Standards solutions for HPLC
Standards solutions for HPLC
Standards solutions for HPLC
Catalog # PERC00890893
Supplier:
PerkinElmer
CAS Number:
Expand image
Standards solutions for HPLC
Catalog # PERC00890893
Supplier:
PerkinElmer
CAS Number:
Specifications
Pk:
5
Description:
Universal test mix for Reversed-phase, 5 ml
Seq:
01
Environmentally Preferable:
Specifications
About this item
Product Details
Product Details