Order Entry
Poland
ContactUsLinkComponent
Anti-Maltose Binding Protein Tag Rabbit Polyclonal Antibody
Anti-Maltose Binding Protein Tag Rabbit Polyclonal Antibody
Catalog # BSENR-1313-100
Supplier:  Biosensis
undefined
Anti-Maltose Binding Protein Tag Rabbit Polyclonal Antibody
Catalog # BSENR-1313-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Maltose Binding Protein Tag
  • Antigen symbol:
    MBP-Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    MBP epitope
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    MBP epitope tag recombinant protein.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Maltose binding protein (MBP) is encoded by the malE gene of E.coli. MBP is often used in protein expression studies because it creates a stable fusion product that does not appear to interfere with the bioactivity of the protein of interest. It also allows for its easy purification from bacterial extracts under mild conditions.

This antibody recognizes the MBP epitope tag fused to the amino- or carboxy- termini of targeted proteins.
Application Information:
Western Blotting (WB) and ELISA. Suggested starting dilutions for WB of 1:1,000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: Maltose Binding Protein Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: MBP epitope
Host: Rabbit
Isotype:
Reactivity: