Anti-CHGA Mouse Monoclonal Antibody [clone: LK2H10]
Catalog # ABNOMAB11338
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:Chromogranin A
- Antigen symbol:CHGA
- Clonality:Monoclonal
- Clone:LK2H10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1113
- Antigen synonyms:CGA
- Storage buffer:PBS, pH 7,4 (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human phaeochromocytoma.
- Purification:Protein A/G affinity chromatography
- Size:100 µg
- Pk:100 µg
Dane techniczne
Informacje o tym produkcie
Mouse monoclonal antibody raised against CHGA.
Recommended Dilutions: Immunohistochemistry: 1:200-1:400 for 30 min at RT; Western Blot: 1:400-1:800
Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: LK2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Pig, Rat