Order Entry
Poland
ContactUsLinkComponent
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: HL-39]
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: HL-39]
Catalog # ABNOMAB0902
Supplier:  Abnova
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: HL-39]
Catalog # ABNOMAB0902
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Major histocompatibility Complex, class II, DR alpha
  • Antigen symbol:
    HLA-DRA
  • Clonality:
    Monoclonal
  • Clone:
    HL-39
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG3
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3122|3123|3125|3126|3127
  • Antigen synonyms:
    MLRW|HLA-DRA1
  • Storage buffer:
    TBS, pH 8.0 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    Native purified HLA-DR from Raji human Burkitt's lymphoma cell line.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native HLA-DR.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: HL-39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human