Order Entry
Poland
ContactUsLinkComponent
Anti-AFP Mouse Monoclonal Antibody [clone: C3]
Anti-AFP Mouse Monoclonal Antibody [clone: C3]
  ABNOMAB1822
 :  Abnova
Anti-AFP Mouse Monoclonal Antibody [clone: C3]
  ABNOMAB1822
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Alpha 1 Fetoprotein Receptor
  • Antigen symbol:
    AFP
  • Clonality:
    Monoclonal
  • Clone:
    C3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Human,
    Dog
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    174
  • Antigen synonyms:
    FETA|Alpha-1-Fetoprotein|HPAFP
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Human AFP.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against AFP.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Pig