Order Entry
Poland
ContactUsLinkComponent
Anti-H2-AB1 Mouse Monoclonal Antibody [clone: IA-C6]
Anti-H2-AB1 Mouse Monoclonal Antibody [clone: IA-C6]
Catalog # ABNOMAB1330
Supplier:  Abnova
Anti-H2-AB1 Mouse Monoclonal Antibody [clone: IA-C6]
Catalog # ABNOMAB1330
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    histocompatibility 2, class II antigen A, beta 1
  • Antigen symbol:
    H2-AB1
  • Clonality:
    Monoclonal
  • Clone:
    IA-C6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    14961
  • Antigen synonyms:
    H-2Ab|Abeta|Ia-2|Rmcs1|IAb|H2-Ab|Ia2|AI845868|I-Abeta
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified from mouse lymph node B cells.
  • Size:
    200 μg
  • Pk:
    200 µg

Specifications

About this item

Mouse monoclonal antibody raised against native H2-Ab1.

Type: Primary
Antigen: H2-AB1
Clonality: Monoclonal
Clone: IA-C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse