Order Entry
Poland
ContactUsLinkComponent
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Catalog # ABNOMAB1474
Supplier:  Abnova
Anti-CDKN1B Mouse Monoclonal Antibody [clone: IMD-27]
Catalog # ABNOMAB1474
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase inhibitor 1B
  • Antigen symbol:
    CDKN1B
  • Clonality:
    Monoclonal
  • Clone:
    IMD-27
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rodent,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length rodent Cdkn1b.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant Cdkn1b.

Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: IMD-27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rodents