IgG2a Isotype Control [clone: PPV-04]
Catalog # ABNOMAB10152
Supplier: Abnova
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:PPV-04
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2a
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, 300 mM NaCl (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a plant pathogen.
Recommended Dilutions: Flow Cytometry: 10 ?g/ml
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: PPV-04
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: