IgG2b Isotype Control [clone: PLRV219]
ABNOMAB10154
: Abnova
- Antibody type:Control
- Clonality:Monoclonal
- Clone:PLRV219
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2b
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against a plant pathogen.
Recommended Dilutions: Flow Cytometry: 10 ?g/ml
Type: Control
Antigen: IgG2 β
Clonality: Monoclonal
Clone: PLRV219
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity: