Order Entry
Poland
ContactUsLinkComponent
IgM Isotype Control [clone: PFR-03]
IgM Isotype Control [clone: PFR-03]
Catalog # ABNOMAB10155
Supplier:  Abnova
IgM Isotype Control [clone: PFR-03]
Catalog # ABNOMAB10155
Supplier:  Abnova

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    PFR-03
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Isotype:
    IgM
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Purification:
    Precipitation method
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a plant pathogen.

Type: Control
Antigen: IgM
Clonality: Monoclonal
Clone: PFR-03
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgM
Reactivity: