Anti-HMGB1 Mouse Monoclonal Antibody [clone: J2E1]
Catalog # ABNOMAB1771
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Box 1
- Antigen symbol:HMGB1
- Clonality:Monoclonal
- Clone:J2E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3146
- Antigen synonyms:high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
- Amino acid number:1 to 215
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-215 of human HMGB1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant HMGB1.
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:100-1:300
Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: J2E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human