Order Entry
Poland
ContactUsLinkComponent
Anti-RAB11FIP3 Rabbit Polyclonal Antibody
Anti-RAB11FIP3 Rabbit Polyclonal Antibody
  ABNOPAB22195
 :  Abnova
Anti-RAB11FIP3 Rabbit Polyclonal Antibody
  ABNOPAB22195
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    RAB11 Family interacting Protein 3
  • Antigen symbol:
    RAB11FIP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9727
  • Antigen synonyms:
    Rab11-FIP3|arfophilin-1|PAC196A12.1|eferin|rab11-family interacting protein 3|FIP3-Rab11|MU-MB-17.148|CART1|EF hands-containing Rab-interacting protein|cytoplasmic adaptor for RAR and TR|rab11 family-interacting protein 3
  • Storage buffer:
    PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    PLFSWTEEPEECGPASCPESAPFRLQGSSSSHRARGEVDVFSPFPAPTAGELALEQGPGSPPQPSDLSQTHPLPSEPVGSQ
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human RAB11FIP3.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against recombinant RAB11FIP3.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RAB11FIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human