Anti-WRNIP1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10242
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Werner Helicase Interacting Protein 1
- Antigen symbol:WRNIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:56897
- Antigen synonyms:bA420G6.2|WHIP
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of human WRNIP1.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of WRNIP1.
Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: WRNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat