Order Entry
Poland
ContactUsLinkComponent
Anti-SLC6A9 Rabbit Polyclonal Antibody
Anti-SLC6A9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB20681
Supplier:  Abnova
Anti-SLC6A9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB20681
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 6 (Neurotransmitter Transporter), Member 9
  • Antigen symbol:
    Solute carrier family 6, member 9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6536
  • Antigen synonyms:
    GlyT-1GlyT1|Glyt 1|SC6A9_HUMAN|Sodium- and chloride-dependent glycine transporter 1|glycine transporter 1|sodium and chloride dependent glycine transporter 1|Solute carrier family 6 member 9.|SLC6A9
  • Storage buffer:
    PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    YCNNPWNTHDCAGVLDASNLTNGSRPAALPSNLSHLLNHSLQRTSPSEEYWRLYVLKLSDDIGNFGE
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human SLC6A9.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant SLC6A9.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: SLC6A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human