Złóż zamówienie
Poland
ContactUsLinkComponent
Anti-CERS5 Rabbit Polyclonal Antibody
Anti-CERS5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13439
Supplier:  Abnova
Anti-CERS5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13439
Supplier:  Abnova

Specifications

  • Typ przeciwciała:
    Primary
  • Nazwa antygenu:
    Ceramide synthase 5
  • Symbol antygenu:
    CERS5
  • Klonalność:
    Poliklonalne
  • Koniugacja:
    Unconjugated
  • Gospodarz:
    Rabbit
  • ImmunoFluorescencja:
    Yes
  • Reaktywność:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Adsorpcja krzyżowa:
    No
  • Format:
    Liquid
  • Identyfikator genu:
    91012
  • Synonimy antygenów:
    LASS5|Trh4
  • Bufor do przechowywania:
    PBS (0,02% sodium azide)
  • Temperatura przechowywania:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to C-terminus 17 amino acids of human LASS5.
  • Rozmiar:
    100 μg
  • Ilość:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of LASS5.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CERS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat