Order Entry
Poland
ContactUsLinkComponent
Anti-Clostridium botulinum Toxin B Mouse Monoclonal Antibody [clone: B362M]
Anti-Clostridium botulinum Toxin B Mouse Monoclonal Antibody [clone: B362M]
Catalog # ABNOMAB4096
Supplier:  Abnova
Anti-Clostridium botulinum Toxin B Mouse Monoclonal Antibody [clone: B362M]
Catalog # ABNOMAB4096
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Clostridium botulinum Toxin B
  • Clonality:
    Monoclonal
  • Clone:
    B362M
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Bacteria
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Culture Supernatant
  • Amino acid number:
    1278 to 1291
  • Storage buffer:
    culture supernatant
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 1278-1291 of Clostridium botulinum Toxin B.
  • Size:
    1 mL
  • Pk:
    1 ml

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin B.

Recommended Dilutions: Immunocytochemistry: 1:25

Type: Primary
Antigen: Clostridium botulinum Toxin B
Clonality: Monoclonal
Clone: B362M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bacteria