Order Entry
Poland
ContactUsLinkComponent
Anti-TGFB1 Mouse Monoclonal Antibody [clone: TB21]
Anti-TGFB1 Mouse Monoclonal Antibody [clone: TB21]
Catalog # ABNOMAB2505
Supplier:  Abnova
Anti-TGFB1 Mouse Monoclonal Antibody [clone: TB21]
Catalog # ABNOMAB2505
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transforming Growth Factor Beta 1
  • Antigen symbol:
    TGFB1
  • Clonality:
    Monoclonal
  • Clone:
    TB21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    7040
  • Antigen synonyms:
    CED|TGFbeta|TGFB|DPD1|LAP
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified human TGFB1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native TGFB1.

Recommended Dilutions: Western Blot: 5-20 ng/ml; Immunohistochemistry: 1:1000

Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: TB21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat