Anti-CFB Mouse Monoclonal Antibody [clone: KT24]
ABNOMAB4954
: Abnova
- Antibody type:Primary
- Antigen name:Complement Factor B
- Antigen symbol:CFB
- Clonality:Monoclonal
- Clone:KT24
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:629
- Antigen synonyms:BFD|CFAB|AHUS4|FB|CFBD|GBG|H2-Bf|BF|PBF2|FBI12|ARMD14
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human IL1R1.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native CFB
Recommended Dilutions: Sandwich ELISA: 1:1000
Type: Primary
Antigen: CFB
Clonality: Monoclonal
Clone: KT24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human