Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]
Catalog # ABNOMAB2525
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Thy1 Cell Surface Antigen
- Antigen symbol:THY1
- Clonality:Monoclonal
- Clone:F15-42-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Cross adsorption:No
- Format:Liquid
- Gene ID:7070
- Antigen synonyms:Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
- Storage buffer:PBS, pH 7,4 (0,09% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human THY1.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against native THY1.
Recommended Dilutions: Flow Cytometry: 10?l of the suggested working dilution 1:10-1:50 to label 10^6 cells in 100?l
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat