Anti-Collagen Type VI Rabbit Polyclonal Antibody
Catalog # ABNOPAB13510
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Collagen Type VI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:phosphate buffer
- Storage temperature:Store at –20 °C. Aliquot after reconstitution to avoid repeated freezing and thawing.
- Immunogen:Native purified Collagen Type VI from human placenta.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against native Collagen Type VI.
Recommended Dilutions: ELISA: 1:200; Immunohistochemistry: 1:400-1:1000; Immunofluorescence: 1:80
Type: Primary
Antigen: Collagen Type VI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human