Anti-APP Rabbit Polyclonal Antibody
Catalog # ABNOPAB13588
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Form:Serum
- Gene ID:351
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Amino acid number:1 to 42
- Sequence:DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with sterile water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-42 of human Amyloid-beta.
- Tested applications:Dot Blot
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Beta amyloid.
- Recommended Dilutions: ELISA: 1:3000; Dot Blot: 1:1000; Western Blot: 1:1000
Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human